missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Galactose, >99%

Catalog No. AAJ15995A7
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ15995A7 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ15995A7 Supplier Thermo Fisher Scientific Supplier No. J15995A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59-23-4 | C6H12O6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00151230

Specifications

Melting Point 164°C to 168°C
Density 1.5
Packaging Plastic bottle
Quantity 5 kg
Beilstein 1724619
Specific Gravity 1.50
Formula Weight 180.16
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D-(+)-Galactose
EINECSNumber 200-416-4
RTECSNumber LW5490000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only