missing translation for 'onlineSavingsMsg'
Learn More

D-Galactose (White to Off-white Powder), Fisher BioReagents

Catalog No. BP656500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP656500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP656500 Supplier Fisher BioReagents Supplier No. BP656 500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D-Galactose is suitable for use in cell culture systems requiring sugar additives.

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6

Specifications

Alpha Vector Galactose
Melting Point 167°C
Color White
Assay Percent Range ≥98 %
Packaging Poly Bottle
Quantity 500 g
Merck Index 15, 4365
Specific Rotation 78.2 to 82.2° (+ or -)
Formula Weight 180.08
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material D-Galactose
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT