accessibility menu, dialog, popup

UserName

CH3OOCH3OCH3OOH

CAS RN 58186-27-9

IUPAC Name: 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Synonyms: idebenone hydroxydecyl ubiquinone 艾地苯醌 idebenone (substance) idebenona idebenone (product) idebenonum
Molecular Weight (g/mol): 338.44
Molecular Formula: C19H30O5
InChi Key: JGPMMRGNQUBGND-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O
Physical Form 
  • (3)
  • (4)
Grade 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)