missing translation for 'onlineSavingsMsg'
Learn More

Idebenone, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014747
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014747 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014747 Supplier Merck Emd Millipore Supplier No. 6780510MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 58186-27-9
Physical Form Neat
Linear Formula C19H30O5
MDL Number MFCD00274552
Type Analytical Standard
Quantity 10 mg
Formula Weight 338.44
Percent Purity ≥97% (HPLC)
Grade Analytical Standard