missing translation for 'onlineSavingsMsg'
Learn More

Idebenone, 98.5%, Spectrum™ Chemical

Catalog No. 18611010
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611010 5 g Amber Glass Bottle
18611009 25 g Amber Glass Bottle
18611008 100 g Amber Glass Bottle
18611007 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611010 Supplier Spectrum Chemical Mfg Cor Supplier No. I20165GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58186-27-9
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.44
InChI Key JGPMMRGNQUBGND-UHFFFAOYSA-N
IUPAC Name 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
SMILES COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O

Specifications

CAS Max % 1
Melting Point 52°C to 55°C
Loss on Drying 0.02
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.985
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Idebenone

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.