missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Idebenone

Catalog No. AC466770050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466770050 5 g
AC466770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466770050 Supplier Thermo Scientific Chemicals Supplier No. 466770050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58186-27-9 | C19H30O5, C19H30O5 | 338.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58186-27-9
Molecular Formula C19H30O5

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: insoluble. Other solubilities: slightly soluble in methanol
Melting Point 52.0°C to 55.0°C
Formula Weight 338.44
MDL Number MFCD00274552
InChI Key JGPMMRGNQUBGND-UHFFFAOYSA-N
SMILES COC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O
IUPAC Name 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 338.44
Chemical Name or Material Idebenone

RUO – Research Use Only