accessibility menu, dialog, popup

UserName

CAS RN 573-58-0

IUPAC Name: disodium 4-amino-3-(2-{4'-[2-(1-amino-4-sulfonatonaphthalen-2-yl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}diazen-1-yl)naphthalene-1-sulfonate
Synonyms: congo red direct red 28 刚果红 kongorot
Molecular Weight (g/mol): 696.66
Molecular Formula: C32H22N6Na2O6S2
InChi Key: IQFVPQOLBLOTPF-UHFFFAOYSA-L
SMILES: [Na+].[Na+].NC1=C2C=CC=CC2=C(C=C1N=NC1=CC=C(C=C1)C1=CC=C(C=C1)N=NC1=CC(=C2C=CC=CC2=C1N)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
Grade 
  • (2)
  • (8)
  • (6)
Color 
  • (1)
  • (1)
  • (6)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (10)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (5)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (8)
  • (6)
  • (1)
  • (1)
  • (6)
  • (4)
  • (1)
  • (10)
  • (1)
  • (2)
  • (1)