missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Congo Red Indicator, 0.1% (w/v), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Congo Red Indicator |
| CAS | 573-58-0, 7732-18-5 |
| CAS Min % | 98.0, 0.1 |
| CAS Max % | 100.0, 0.1 |
| Concentration | 0.1% (w/v) |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Color | Red |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Show More |