missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Congo Red, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Packaging | Pouch |
| Color | Brown/Red |
| pH | 8 to 9.5 |
| Quantity | 10 g |
| Physical Form | Powder |
| Grade | Indicator |
| Chemical Name or Material | Congo Red |