missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Congo Red, indicator grade

Catalog No. AAB2431014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2431014 25 g
AAB2431022 100 g
AAB2431036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2431014 Supplier Thermo Scientific Chemicals Supplier No. B2431014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 573-58-0 | C32H22N6Na2O6S2 | 696.664 g/mol

Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

Solubility
Soluble in water 25 g/L.

Notes
Store away from oxidizing agents, acids, reducing agents.

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
MDL Number MFCD00004028
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym C.I. 22120
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >360°C
Beilstein 3894858
Merck Index 14,2498
Solubility Information Soluble in water 25g/L.
Formula Weight 696.67
Grade Indicator
Odor Odorless
Quantity 25 g
Chemical Name or Material Congo Red
Hazard Category H350-H361d
Hazard Statement GHS H Statement
H350-H361
May cause cancer.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 209-358-4
RTECSNumber QK1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only