missing translation for 'onlineSavingsMsg'
Learn More

Congo Red 98.0+%, TCI America™

Catalog No. C055025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C055025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C055025G Supplier TCI America Supplier No. C055025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
MDL Number MFCD00004028
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym Direct Red 28
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 696.66
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Congo Red
RTECSNumber QK1400000
TSCA Yes