missing translation for 'onlineSavingsMsg'
Learn More

Congo Red Indicator, 0.1% (w/v), pH 3.0 to 5.0 Blue to Red, Certified, LabChem™

Catalog No. LC133607
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC133607 125 mL
LC133601 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC133607 Supplier LabChem Supplier No. LC133607

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 573-58-0 , 7732-18-5
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

CAS Max % 99.9, 0.1
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Linear Formula C32H22N6O6S2Na2
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Density 1g/mL
Color Red
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Formula Weight 696.67
Grade Certified
Chemical Name or Material Congo Red Indicator, 0.1% (w/v), pH 3.0 to 5.0 Blue to Red
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause cancer.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
If exposed or concerned: Get medical advice/attention.
Recommended Storage Room Temperature