accessibility menu, dialog, popup

UserName

H3C(S)OHONH2

CAS RN 56-41-7

IUPAC Name: (2S)-2-aminopropanoic acid
Synonyms: L-alanine|L alanine|alanine, L-|Ala|2-aminopropanoic acid|D,L-alanine|L-alanin|2-aminopropionic acid|alaninum|delta-alanine
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: QNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILES: C[C@H](N)C(O)=O
Quantity 
  • (4)
  • (6)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (8)
  • (9)
  • (1)
Grade 
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (17)
  • (2)
  • (1)
  • (3)
  • (8)
  • (2)
  • (12)
  • (4)
  • (6)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (8)
  • (9)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)