missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, 99%, MP Biomedicals™

Catalog No. ICN1002873
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1002873 25 g
ICN10028701 1 kg
ICN10028705 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN1002873 Supplier MP Biomedicals Inc Supplier No. 02100287.3

A nonessential amino acid that plays a key role in the glucose-alanine cycle between muscle tissue and the liver. In amino acid-degrading tissues such as muscle, amino groups are pooled as glutamate by transamination reactions

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 0.99
Density 1.40g/mL
Quantity 25 g
Formula Weight 89.1
Percent Purity ≥99%
Grade Biochemical
Physical Form Powder
Chemical Name or Material L-Alanine
EINECSNumber 200-273-8