missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine 99.0+%, TCI America™

Catalog No. A017925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A017925G 25 g
A0179250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A017925G Supplier TCI America Supplier No. A017925G

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Melting Point 315°C
Color White
Quantity 25 g
Formula Weight 89.09
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine
EINECSNumber (9)-1553
RTECSNumber AY2990000
TSCA Yes