missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Alanine, Cell Culture Reagent

Catalog No. AAJ6027936
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6027936 500 g
AAJ6027918 50 g
AAJ6027922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6027936 Supplier Thermo Scientific Chemicals Supplier No. J6027936

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol

It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

Solubility
Soluble in water 166.5 g/L..

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Melting Point ∽295°C (decomposition)
Color White
Beilstein 1720248
Merck Index 14,204
Solubility Information Soluble in water 166.5g/L..
Quantity 500 g
Formula Weight 89.09
Optical Rotation +14° (c=5 in 5N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Alanine
Show More Show Less
EINECSNumber 200-273-8
RTECSNumber AY2990000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only