missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014640
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014640 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014640 Supplier Merck Emd Millipore Supplier No. PHR11101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-Alanine, a non-essential amino acid, is produced enzymatically from L-aspartate using aspartate beta-decarboxylase. It is the smallest among all the amino acids.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 56-41-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula C3H7 NO2
MDL Number MFCD00064410
Synonym (S)-2-Aminopropionic acid; L-alpha-Aminopropionic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 89.09
Grade Certified Reference Material