missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Alanine, ≥98%, Calbiochem™,

Catalog No. 1250100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1250100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1250100GM Supplier MilliporeSigma™ Supplier No. 1250100GM

Non-essential amino acid supplied as a white solid for laboratory applications.

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

pH 5.5 to 7.0
Color White
Packaging Glass Bottle
Quantity 100 g
Percent Purity ≥98%
Grade Biochemical
Physical Form Solid