CAS RN 50-70-4
Sorbitol, Fisher BioReagents
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.172 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
Thermo Scientific Chemicals D-Sorbitol, 97%
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 MDL Number: MFCD00004708 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
Sorbitol, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards
Thermo Scientific Chemicals D-Sorbitol, 98%
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.172 MDL Number: MFCD00004708 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
D-Sorbitol 97.0+%, TCI America™
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.172 MDL Number: MFCD00004708 InChI Key: FBPFZTCFMRRESA-JGWLITMVSA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol PubChem CID: 5780 ChEBI: CHEBI:17924 IUPAC Name: (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol SMILES: C(C(C(C(C(CO)O)O)O)O)O
D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98%, MP Biomedicals™
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N Synonym: d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,glucarine,diakarmon,multitol,sorbilande PubChem CID: 5780 ChEBI: CHEBI:17924
Sorbitol Solution, USP, 70%, Spectrum™ Chemical
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Sorbitol, Powder, NF, 91-100.5%, Spectrum™ Chemical
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Sorbitol, Powder, FCC, 91-100.5%, Spectrum™ Chemical
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Sorbitol Solution, USP, 70%, Spectrum™ Chemical
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Sorbitol, Powder, NF, 91-100.5%, Spectrum™ Chemical
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Sorbitol Solution, FCC, 70%, Spectrum™ Chemical
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Sorbitol, Powder, FCC, 91-100.5%, Spectrum™ Chemical
CAS: 50-70-4 Molecular Formula: C6H14O6 Molecular Weight (g/mol): 182.17 InChI Key: FBPFZTCFMRRESA-UHFFFAOYNA-N IUPAC Name: hexane-1,2,3,4,5,6-hexol SMILES: OCC(O)C(O)C(O)C(O)CO
Medisca Sorbitol NF
Sorbitol NF can be white granules, powder, or crystalline masses. It is odorless and hygroscopic. Alternative name(s): d-Sorbitol