missing translation for 'onlineSavingsMsg'
Learn More

Medisca Sorbitol NF

Catalog No. 14185248
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185248 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185248 Supplier Medisca Supplier No. 052708
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sorbitol NF can be white granules, powder, or crystalline masses. It is odorless and hygroscopic. Alternative name(s): d-Sorbitol

Sorbitol NF can be white granules, powder, or crystalline masses. It is odorless and hygroscopic.

Alternative name(s): d-Sorbitol

Specifications

Chemical Name or Material Sorbitol NF
Packaging Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 50-70-4
Type Drug Standard
Molecular Weight (g/mol) 182.17
Quantity 500 g
Percent Purity 91 to 100.5 %
Grade NF