missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111014375
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014375 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014375 Supplier Merck Emd Millipore Supplier No. PHR10061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 50-70-4
Vapor Pressure <0.1 mmHg (25°C)
Vapor Density <1 (vs. air)
Physical Form Neat
Linear Formula C6H14O6
MDL Number MFCD00004708
Melting Point 98°C to 100°C
Synonym D-Sorbitol; D-Glucitol
Quantity 1 g
Formula Weight 182.17
Grade Certified Reference Material
Show More Show Less