missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, Fisher BioReagents

Catalog No. BP439500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP439500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP439500 Supplier Fisher BioReagents Supplier No. FLBP439 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sorbitol is used in the preparation of isoelectric focusing gels.

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 75°C
Color White
Density 1.489g/cm³
pH 6 to 7
Boiling Point 295°C
Assay Percent Range 91.0 to 100.5 %
Packaging Poly Bottle
Quantity 500 g
Merck Index 15, 8851
Formula Weight 182.17
Percent Purity 91.0 to 100.5%
Physical Form Solid
Chemical Name or Material Sorbitol
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT