accessibility menu, dialog, popup

UserName

CAS RN 4595-60-2

IUPAC Name: 2-bromopyrimidine
Synonyms: pyrimidine, 2-bromo-
Molecular Weight (g/mol): 158.99
Molecular Formula: C4H3BrN2
InChi Key: PGFIHORVILKHIA-UHFFFAOYSA-N
SMILES: BrC1=NC=CC=N1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)