missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyrimidine 98.0+%, TCI America™

Catalog No. B28325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28325G 5 g
B283225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28325G Supplier TCI America Supplier No. B28325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4595-60-2
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 158.99
MDL Number MFCD00014601
InChI Key PGFIHORVILKHIA-UHFFFAOYSA-N
Synonym pyrimidine, 2-bromo, 2-brompyrimidin, bromopyrimidine, 2-bromo pyrimidine, 2-bromo-pyrimidine, 2-bromanylpyrimidine, pubchem6901, acmc-1aqvo, 2-bromo-1,3-diazine, 2-bromopyrimidine
PubChem CID 78345
IUPAC Name 2-bromopyrimidine
SMILES BrC1=NC=CC=N1

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 64°C
Quantity 5 g
Formula Weight 158.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromopyrimidine
TSCA Yes
Recommended Storage Refrigerator