missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyrimidine, 98+%

Catalog No. AAA1379122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1379122 100 g
AAA1379106 5 g
AAA1379114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1379122 Supplier Thermo Scientific Chemicals Supplier No. A1379122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4595-60-2 | C4H3BrN2 | 158.99 g/mol

2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, acid chlorides.

Chemical Identifiers

CAS 4595-60-2
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 158.99
MDL Number MFCD00014601
InChI Key PGFIHORVILKHIA-UHFFFAOYSA-N
Synonym pyrimidine, 2-bromo, 2-brompyrimidin, bromopyrimidine, 2-bromo pyrimidine, 2-bromo-pyrimidine, 2-bromanylpyrimidine, pubchem6901, acmc-1aqvo, 2-bromo-1,3-diazine, 2-bromopyrimidine
PubChem CID 78345
IUPAC Name 2-bromopyrimidine
SMILES BrC1=NC=CC=N1

Specifications

Melting Point 53°C to 57°C
Boiling Point 62°C to 64°C (1.5 mmHg)
Quantity 100 g
Beilstein 605945
Solubility Information Miscible with water.
Formula Weight 158.99
Percent Purity ≥98%
Chemical Name or Material 2-Bromopyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-993-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only