missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyrimidine, 97+%

Catalog No. AC403230050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403230050 5 g Glass bottle
AC403230010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403230050 Supplier Thermo Scientific Chemicals Supplier No. 403230050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4595-60-2 | C4H3BrN2 | 158.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4595-60-2
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 158.99
MDL Number MFCD00014601
InChI Key PGFIHORVILKHIA-UHFFFAOYSA-N
Synonym pyrimidine, 2-bromo, 2-brompyrimidin, bromopyrimidine, 2-bromo pyrimidine, 2-bromo-pyrimidine, 2-bromanylpyrimidine, pubchem6901, acmc-1aqvo, 2-bromo-1,3-diazine, 2-bromopyrimidine
PubChem CID 78345
IUPAC Name 2-bromopyrimidine
SMILES BrC1=NC=CC=N1

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 55.0°C to 56.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 158.99
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromopyrimidine
Show More Show Less
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 224-993-7

RUO – Research Use Only