accessibility menu, dialog, popup

UserName

CAS RN 403811-55-2

IUPAC Name: 5-[(4-ethylphenyl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: 5-[(4-ethylphenyl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 249.35
Molecular Formula: C12H11NOS2
InChi Key: SVXDHPADAXBMFB-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C2SC(=S)NC2=O)C=C1
Color 
  • (2)
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results