missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ 10058-F4
Description
10058-F4 is a cell permeable c-Myc-Max dimerization inhibitor. Inhibits proliferation, induces apoptosis and arrests cells in G0/G1 in rat1a-c-Myc cells. Also reduces tumor growth in vivo. In lymphoma cells, 10058-F4 decreases glucose uptake and expression of glycolysis-associated genes and reduces cell viability.
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Formula Weight | 249.35 |
| Physical Form | Solid |
| Color | Yellow |
| Purity | >99% |
| Molecular Weight (g/mol) | 249.346 |
| Chemical Name or Material | 10058-F4 |
Safety and Handling
| Recommended Storage | Store at +4°C |