missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ c-Myc Inhibitor

Catalog No. 47595610MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47595610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47595610MG Supplier MilliporeSigma™ Supplier No. 47595610MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The c-Myc Inhibitor, also referenced under CAS 403811-55-2, controls the biological activity of c-Myc.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color Yellow
Description 95% by HPLC (sum of two isomers)
Product Type Inhibitor
Target Proteins c-Myc
CAS 403811-55-2
Molecular Formula C12H11NOS2
Formula Weight 249.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Show More Show Less