accessibility menu, dialog, popup

UserName

H3CNCH3FPFFFFF

CAS RN 401788-99-6

IUPAC Name: 1-butyl-4-methylpyridin-1-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-butyl-4-methylpyridin-1-ium hexafluorophosphate
Molecular Weight (g/mol): 295.21
Molecular Formula: C10H16F6NP
InChi Key: XBSZSSGZQFAUPF-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCC[N+]1=CC=C(C)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)