missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-4-methylpyridinium hexafluorophosphate, 99%

Catalog No. AAH2735518
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2735518 50 g
AAH2735506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2735518 Supplier Thermo Scientific Chemicals Supplier No. H2735518
Only null left

CAS: 401788-99-6 | C10H16F6NP | 295.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 401788-99-6
Molecular Formula C10H16F6NP
Molecular Weight (g/mol) 295.209
MDL Number MFCD03095438
InChI Key XBSZSSGZQFAUPF-UHFFFAOYSA-N
Synonym 1-butyl-4-methylpyridinium hexafluorophosphate, 4mbppf6, 1-butyl-4-methylpyridin-1-ium hexafluorophosphate, acmc-209jbi, dsstox_cid_29152, dsstox_rid_83371, dsstox_gsid_49296
PubChem CID 11254899
IUPAC Name 1-butyl-4-methylpyridin-1-ium;hexafluorophosphate
SMILES CCCC[N+]1=CC=C(C=C1)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 45°C
Quantity 50 g
Formula Weight 295.22
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-4-methylpyridinium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only