missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-4-methylpyridinium Hexafluorophosphate 97.0+%, TCI America™

Catalog No. B27015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27015G 5 g
B270125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27015G Supplier TCI America Supplier No. B27015G
Only null left

Chemical Identifiers

CAS 401788-99-6
Molecular Formula C10H16F6NP
Molecular Weight (g/mol) 295.209
InChI Key XBSZSSGZQFAUPF-UHFFFAOYSA-N
Synonym 1-butyl-4-methylpyridinium hexafluorophosphate, 4mbppf6, 1-butyl-4-methylpyridin-1-ium hexafluorophosphate, acmc-209jbi, dsstox_cid_29152, dsstox_rid_83371, dsstox_gsid_49296
PubChem CID 11254899
IUPAC Name 1-butyl-4-methylpyridin-1-ium;hexafluorophosphate
SMILES CCCC[N+]1=CC=C(C=C1)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 42°C
Quantity 5 g
Formula Weight 295.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-4-methylpyridinium Hexafluorophosphate