CAS RN 38966-21-1
IUPAC Name:
(1S,2S,5R,6R,7R,10S,12R,13R)-6,13-bis(hydroxymethyl)-2,6-dimethyltetracyclo[10.3.1.0¹,¹⁰.0²,⁷]hexadecane-5,13-diol
Synonyms:
aphidicolin
Molecular Weight (g/mol):
338.49
Molecular Formula:
C20H34O4
InChi Key:
NOFOAYPPHIUXJR-APNQCZIXSA-N
SMILES:
C[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H]1CC[C@H]1C[C@@H]3C[C@]21CC[C@]3(O)CO
1
–
4
of
4
results
1
MilliporeSigma™ Calbiochem™ Aphidicolin
Aphidicolin, CAS 38966-21-1, is a cell-permeable antibiotic that acts as a cell synchronization agent. Blocks the cell cycle at early S-phase.
2
Thermo Scientific Chemicals Aphidicolin
CAS: 38966-21-1 Molecular Formula: C20H34O4 Synonym: APC
3
Aphidicolin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Aphidicolin is a diterpenoid metabolite of the fungi, Cephalosporium aphidicola and Phoma betae. It is known to be an antiviral drug and inhibits the incorporation of thymidine into DNA of cultured human embryonic lung cells.
4
R&D Systems™ Aphidicolin
A Tocris product, part of R&D Systems. DNA polymerase α, δ and ε inhibitor