missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Aphidicolin
Description
Aphidicolin is a DNA polymerase α, δ and ε inhibitor. Exhibits selectivity over DNA polymerase β and γ. Antimitotic, antibiotic and antiviral.
Specifications
Specifications
| Chemical Name or Material | (3 R,4 R,4 aR,6 AS,8 R,9 R,11 AS,11 bS)-Tetradecahydro-3,9-dihydroxy-4,11 b-dimethyl-8,11 A-methano-11 aH-cyclohepta[a]naphthalene-4,9-dimethanol |
| CAS | 38966-21-1 |
| Quantity | 1 mg |
| Product Line | Tocris |
| Target | RNA/DNA Polymerase Inhibitors |
| Molecular Formula | C20H34O4 |
| Formula Weight | 338.48 |
| Purity | 98% |