accessibility menu, dialog, popup

UserName

CAS RN 38748-32-2

IUPAC Name: (1S,2S,4S,5S,7R,8R,9S,11S,13S)-8-hydroxy-1-methyl-7-(propan-2-yl)-3,6,10,16-tetraoxaheptacyclo[11.7.0.0²,⁴.0²,⁹.0⁵,⁷.0⁹,¹¹.0¹⁴,¹⁸]icos-14(18)-en-17-one
Synonyms: triptolide
Molecular Weight (g/mol): 360.41
Molecular Formula: C20H24O6
InChi Key: DFBIRQPKNDILPW-CIVMWXNOSA-N
SMILES: [H][C@@]12C[C@@H]3O[C@@]33[C@H](O)[C@]4(O[C@H]4[C@@H]4O[C@]34[C@@]1(C)CCC1=C2COC1=O)C(C)C
Form 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results