missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Triptolide, Tripterygium wilfordii
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Off-white |
| Product Type | Inhibitor |
| Target Proteins | Activate DEVD-cleaving caspases |
| CAS | 38748-32-2 |
| Molecular Formula | C20H24O6 |
| Formula Weight | 360.4g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 5 mg |
| Show More |