accessibility menu, dialog, popup

UserName

HClH3CNCH3ONHOCl

CAS RN 34552-83-5

IUPAC Name: hydrogen 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide chloride
Synonyms: loperamide
Molecular Weight (g/mol): 513.5
Molecular Formula: C29H34Cl2N2O2
InChi Key: PGYPOBZJRVSMDS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)C(=O)C(CCN1CCC(O)(CC1)C1=CC=C(Cl)C=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)