missing translation for 'onlineSavingsMsg'
Learn More

Loperamide hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019402
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019402 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019402 Supplier Merck Emd Millipore Supplier No. PHR11621G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Loperamide hydrochloride finds application in the regulation and symptomatic relief of long acting diarrhea linked with inflammatory bowel diseases. It can also be used as a muscle relaxant agent.

Specifications

CAS 34552-83-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C29H33 ClN2O2 · HCl
MDL Number MFCD00058581
Synonym 4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinebutyramide hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 513.50
Grade Certified Reference Material