missing translation for 'onlineSavingsMsg'
Learn More

Loperamide hydrochloride, 98+%

Catalog No. AAJ6016814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6016814 25 g
AAJ6016806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6016814 Supplier Thermo Scientific Chemicals Supplier No. J6016814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34552-83-5 | C29H34Cl2N2O2 | 513.503 g/mol

At nanomolar concentrations, binds to mu-opioid receptors. L-type calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • At nanomolar concentrations, binds to mu-opioid receptors
  • L-type calcium channel blocker

Chemical Identifiers

CAS 34552-83-5
Molecular Formula C29H34Cl2N2O2
Molecular Weight (g/mol) 513.503
MDL Number MFCD00058581
InChI Key PGYPOBZJRVSMDS-UHFFFAOYSA-N
Synonym loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
PubChem CID 71420
ChEBI CHEBI:6533
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
SMILES CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl

Specifications

Color White
Quantity 25 g
UN Number UN2811
Merck Index 14,5571
Formula Weight 513.51
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Loperamide hydrochloride
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 252-082-4
RTECSNumber TM4960000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only