missing translation for 'onlineSavingsMsg'
Learn More

Loperamide Hydrochloride 98.0+%, TCI America™

Catalog No. L01545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01545G 5 g
L015425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01545G Supplier TCI America Supplier No. L01545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34552-83-5
Molecular Formula C29H34Cl2N2O2
Molecular Weight (g/mol) 513.503
MDL Number MFCD00058581
InChI Key PGYPOBZJRVSMDS-UHFFFAOYSA-N
Synonym loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
PubChem CID 71420
ChEBI CHEBI:6533
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
SMILES CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl

Specifications

Melting Point 223°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 513.50
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Loperamide Hydrochloride
RTECSNumber TM4960000
TSCA No