missing translation for 'onlineSavingsMsg'
Learn More

Loperamide System Suitability Mixture, MilliporeSigma™ Supelco™

Catalog No. 111002475
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002475 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002475 Supplier Merck Emd Millipore Supplier No. 137001025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Specifications

CAS 34552-83-5
Purity Grade Notes These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Type USP Reference Standard
Quantity 25 mg
Grade Pharmaceutical Primary Standard