accessibility menu, dialog, popup

UserName

H3CNHNOClSH2NOO

CAS RN 26807-65-8

IUPAC Name: 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide
Synonyms: indapamide
Molecular Weight (g/mol): 365.83
Molecular Formula: C16H16ClN3O3S
InChi Key: NDDAHWYSQHTHNT-UHFFFAOYNA-N
SMILES: CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
  • (1)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)