missing translation for 'onlineSavingsMsg'
Learn More

Indapamide 97.0+%, TCI America™

Catalog No. I07305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07305G 5 g
I07301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07305G Supplier TCI America Supplier No. I07305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26807-65-8
Molecular Formula C16H16ClN3O3S
Molecular Weight (g/mol) 365.83
MDL Number MFCD00079375
InChI Key NDDAHWYSQHTHNT-UHFFFAOYNA-N
Synonym indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol
PubChem CID 3702
ChEBI CHEBI:5893
IUPAC Name 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide
SMILES CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 5 g
Formula Weight 365.83
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Indapamide
RTECSNumber CV2451200
TSCA No