missing translation for 'onlineSavingsMsg'
Learn More

Indapamide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013784
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013784 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013784 Supplier Merck Emd Millipore Supplier No. PHR16601G

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Indapamide is a diuretic that is categorized under the family of benzothiadiazines compounds. It is an oral drug that is usually prescribed as an antihypertensive agent for patients with mild to moderate hypertension. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 26807-65-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C16H16 ClN3O3 S
MDL Number MFCD00079375
Synonym Indapamide; N-(4-Chloro-3-sulfamoylbenzamido)-2-methylindoline
Type Certified Reference Material
Quantity 1 g
Formula Weight 365.83
Grade Certified Reference Material