missing translation for 'onlineSavingsMsg'
Learn More

Indapamide, MilliporeSigma™ Supelco™

Catalog No. 111013783
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013783 250 mg
111013782 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013783 Supplier Merck Emd Millipore Supplier No. I1887250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indapamide is a deuretic drug, belonging to the class of benzothiadiazines. It is commonly used for the treatment of hypertension. The drug acts by causing a drop in systolic, diastolic and mean blood pressure.

Featured Industries: Pharmaceutical (small molecule)

Specifications

Chemical Name or Material Indapamide
CAS 26807-65-8
Physical Form Neat
Linear Formula C16H16 ClN3O3 S
Technique Gas Chromatography
MDL Number MFCD00079375
Synonym N-(4-Chloro-3-sulfamoylbenzamido)-2-methylindoline
Type Analytical Standard
Solvent or Matrix Neat
Molecular Weight (g/mol) 365.83
Industry Type Drug Analysis
Quantity 250 mg
Formula Weight 365.83
Grade Analytical Standard
Show More Show Less