CAS RN 25535-16-4
MilliporeSigma™ Propidium Iodide, Calbiochem™
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.406 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
MilliporeSigma™ Propidium Iodide, Calbiochem™,
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.406 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
MilliporeSigma™ Propidium Iodide, Calbiochem™,
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.406 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
MilliporeSigma™ Propidium Iodide Solution, Calbiochem™,
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.406 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
Propidium iodide
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.406 MDL Number: MFCD00011921 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
Propidium iodide, 1mg/ml aqueous soln.
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.406 MDL Number: MFCD00011921 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
Thermo Scientific Chemicals Propidium iodide, 95%
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.39 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
Thermo Scientific Chemicals Propidium iodide, 95%
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.39 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
Propidium Iodide, MP Biomedicals
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.406 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
Propidium Iodide, 99%, For HPLC analysis, MP Biomedicals™
CAS: 25535-16-4 Molecular Formula: C27H34I2N4 Molecular Weight (g/mol): 668.406 InChI Key: XJMOSONTPMZWPB-UHFFFAOYSA-M Synonym: propidium iodide,propidium diiodide,3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide,unii-tp416o228t,3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide,3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide,phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide PubChem CID: 104981 ChEBI: CHEBI:51240 IUPAC Name: 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide SMILES: CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
R&D Systems™ Propidium iodide
A Tocris product, part of R&D Systems. Red-fluorescent DNA stain; membrane impermeant to live cells. Pi staining differentiates live and dead cells