missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Propidium Iodide, Calbiochem™,

Catalog No. 537059250MG
Change view
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
537059250MG 250 mg Glass Bottle
537059100MG 100 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 537059250MG Supplier MilliporeSigma™ Supplier No. 537059250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25535-16-4
Molecular Formula C27H34I2N4
Molecular Weight (g/mol) 668.406
InChI Key XJMOSONTPMZWPB-UHFFFAOYSA-M
Synonym propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
PubChem CID 104981
ChEBI CHEBI:51240
IUPAC Name 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
SMILES CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]

Specifications

Color Red
Packaging Glass Bottle
Quantity 250 mg
Physical Form Solid