missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Propidium iodide, 95%

Catalog No. AC440301000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440301000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440301000 Supplier Thermo Scientific Chemicals Supplier No. 440301000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25535-16-4 | C27H34I2N4 | 668.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25535-16-4
Molecular Formula C27H34I2N4
Molecular Weight (g/mol) 668.39
InChI Key XJMOSONTPMZWPB-UHFFFAOYSA-M
Synonym propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
PubChem CID 104981
ChEBI CHEBI:51240
IUPAC Name 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
SMILES CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]

Specifications

Melting Point 220.0°C to 225.0°C
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: soluble.
Formula Weight 668.39
Percent Purity 95%
Chemical Name or Material Propidium iodide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 247-081-

RUO – Research Use Only