missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Propidium Iodide, Calbiochem™

Catalog No. 5370591GM
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5370591GM 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5370591GM Supplier MilliporeSigma™ Supplier No. 5370591GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Membrane impermeable DNA intercalator. Used to differentiate between apoptotic and necrotic cell death. Useful for staining apoptotic cells and nuclei by flow cytometry.

  • Membrane impermeable DNA intercalator
  • Red fluorescence at 488 nm
  • Useful for staining apoptotic cells and nuclei by flow cytometry
  • Used to differentiate between apoptotic and necrotic cell death
  • Used together with common green fluorescent probes labeled with fluorescein isothiocyanate

Chemical Identifiers

CAS 25535-16-4
Molecular Formula C27H34I2N4
Molecular Weight (g/mol) 668.406
InChI Key XJMOSONTPMZWPB-UHFFFAOYSA-M
Synonym propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
PubChem CID 104981
ChEBI CHEBI:51240
IUPAC Name 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
SMILES CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]

Specifications

Packaging Glass bottle
Quantity 1 g
Physical Form Solid
Hazard Statement R: 36/37/38
Irritating to eyes, respiratory system and skin.
Precautionary Statement S: 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice
RTECSNumber SF7949600
storageNote1 Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C
Recommended Storage 2°C to +8°C