accessibility menu, dialog, popup

UserName

CAS RN 252917-06-9

CH3NHNNNClClHNNHNN

CAS RN 252917-06-9

IUPAC Name: 6-[(2-{[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile
Synonyms: 6-[(2-{[4-(2,4-dichlorophenyl)-5-(4-methyl-3H-imidazol-2-yl)pyrimidin-2-yl]amino}ethyl)amino]pyridine-3-carbonitrile
Molecular Weight (g/mol): 465.34
Molecular Formula: C22H18Cl2N8
InChi Key: AQGNHMOJWBZFQQ-UHFFFAOYSA-N
SMILES: CC1=CN=C(N1)C1=CN=C(NCCNC2=NC=C(C=C2)C#N)N=C1C1=C(Cl)C=C(Cl)C=C1
Form 
  • (1)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)